Assay Details
| Gene Symbol: Tmem218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025464.2 | NP_079740.1 | 2 - 3 | 182 | 72 | |
| GenBank mRNA | AK003010.1 | - | 2 - 3 | 179 | 72 | |
| AK007575.1 | - | 2 - 3 | 72 | |||
| AK008902.1 | - | 2 - 3 | 153 | 72 | ||
| AK161115.1 | - | 184 | 72 | |||
| BC034160.1 | - | 2 - 3 | 182 | 5049687 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem218
Genomic Map