Assay Details
| Gene Symbol: Sln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025540.2 | NP_079816.1 | 1 - 2 | 72 | 63 | |
| GenBank mRNA | AK008863.1 | - | 1 - 2 | 75 | 63 | |
| AK008896.1 | - | 1 - 2 | 73 | 63 | ||
| AK009005.1 | - | 1 - 2 | 75 | 63 | ||
| AK009809.1 | - | 160 | 63 | |||
| BC028496.1 | - | 1 - 2 | 53 | 1397989 | 63 | |
| CA460968.1 | - | 1 - 2 | 110 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008896.1,AK009005.1,AK009809.1,BC028496.1,NM_025540.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | NM_025540.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | AK008863.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | AK008896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | AK009005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | AK009809.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | NM_025540.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | AK008863.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | AK008896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | AK009005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413395 | AK009809.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532019 | AK005862.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532019 | BC049736.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532019 | NM_028825.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79577 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162199 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | NM_025540.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | AK008863.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | AK008896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | AK009005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | AK009809.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | NM_025540.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | AK008863.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | AK008896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | AK009005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413395 | AK009809.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26522815 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162199 | - | - |
| Whole Mouse Genome G4122A | A_52_P413395 | NM_025540.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413395 | AK008863.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413395 | AK008896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413395 | AK009005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413395 | AK009809.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413395 | NM_025540.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413395 | AK008863.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413395 | AK008896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413395 | AK009005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413395 | AK009809.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297498 | AK009809.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114855 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162199 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07819716 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07829390 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07829389 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07829384 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07829397 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07829394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26522781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07911306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07829395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07865361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07819713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07868979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07523920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07480737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07556580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07819716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07868983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07482440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07865366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07868982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26522760 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10585393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585392 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420884_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420883_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10585390 | - | - |
| Mouse Genome 430 2.0 Array | 1420883_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420884_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420884_at | - | - |
| Mouse Genome 430 2.0 Array | 1420883_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66402_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66402_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900004624.mm.1 | NM_025540.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004624.mm.1 | NM_025540.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004624.mm.1 | TC0900000648.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900004625.mm.1 | NM_025540.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004625.mm.1 | NM_025540.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004625.mm.1 | TC0900000648.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420883_PM_at | - | - |
| Mouse Expression Set 430 | 1420884_PM_at | - | - |
| Mouse Expression Set 430 | 1420883_at | - | - |
| Mouse Expression Set 430 | 1420884_at | - | - |
| Murine Genome U74v2 Set | 95048_PM_at | - | - |
| Murine Genome U74v2 Set | 95048_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420883_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420884_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420883_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420884_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420883_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420884_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420883_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420884_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 913347 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2370239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002370239 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2662878 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2662878 | - | - |
| mouse-6 v1.1 (Illumina) | scl36944.2_280-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2370239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36944.2_280-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370239 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2918875 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2918875 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2662878 | - | - |

Sln
Genomic Map