| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800014131.mm.1 |
NM_025556.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014131.mm.1 |
NM_025556.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014131.mm.1 |
TC0800001745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800014133.mm.1 |
NM_025556.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014133.mm.1 |
NM_025556.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014133.mm.1 |
TC0800001745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800014132.mm.1 |
NM_025556.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014132.mm.1 |
NM_025556.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014132.mm.1 |
TC0800001745.mm.1 |
MTA TC |