Assay Details
| Gene Symbol: Sri | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025618.3 | NP_079894.2 | 1 - 2 | 141 | 91 | |
| GenBank mRNA | AK008404.1 | - | 1 - 2 | 93 | 91 | |
| AK008783.1 | - | 1 - 2 | 142 | 91 | ||
| AK008970.1 | - | 1 - 2 | 90 | 91 | ||
| BY708689.1 | - | 1 - 2 | 139 | 91 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008404.1,AK008783.1,AK008970.1,NM_025618.3 transcripts.
Set Membership: |
|

Sri
Genomic Map