Assay Details
| Gene Symbol: Sri | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080974.2 | NP_001074443.1 | 2 - 3 | 178 | 73 | |
| NM_025618.3 | NP_079894.2 | 2 - 3 | 218 | 73 | ||
| GenBank mRNA | AK008404.1 | - | 2 - 3 | 170 | 73 | |
| AK008783.1 | - | 2 - 3 | 219 | 73 | ||
| AK008970.1 | - | 2 - 3 | 167 | 73 | ||
| AK134747.1 | - | 2 - 3 | 180 | 73 | ||
| AK146071.1 | - | 2 - 3 | 175 | 73 | ||
| AK146911.1 | - | 2 - 3 | 159 | 73 | ||
| AK168572.1 | - | 2 - 3 | 159 | 73 | ||
| AK168792.1 | - | 2 - 3 | 159 | 73 | ||
| AK168900.1 | - | 2 - 3 | 150 | 73 | ||
| BC065790.1 | - | 2 - 3 | 145 | 948616 | 73 | |
| BY708689.1 | - | 2 - 3 | 216 | 73 | ||
More Information
Set Membership: |
|

Sri
Genomic Map