Assay Details
| Gene Symbol: Cenpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080158.1 | NP_001073627.1 | 3 - 4 | 381 | 112 | |
| NM_025639.4 | NP_079915.1 | 4 - 5 | 382 | 112 | ||
| GenBank mRNA | AK010885.1 | - | 4 - 5 | 374 | 112 | |
| AK011191.1 | - | 4 - 5 | 359 | 112 | ||
| AK011227.1 | - | 4 - 5 | 366 | 112 | ||
| AK011464.1 | - | 4 - 5 | 383 | 112 | ||
| AK011755.1 | - | 4 - 5 | 375 | 112 | ||
| AK011793.1 | - | 4 - 5 | 368 | 112 | ||
| AK017679.1 | - | 4 - 5 | 375 | 112 | ||
| AY243536.1 | - | 4 - 5 | 343 | 112 | ||
| AY243537.1 | - | 4 - 5 | 343 | 112 | ||
| BC009160.1 | - | 4 - 5 | 361 | 2655787 | 112 | |
| BG276334.1 | - | 3 - 4 | 381 | 112 | ||
| BY710734.1 | - | 4 - 5 | 383 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpm
Genomic Map