Assay Details
| Gene Symbol: Rdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025654.2 | NP_079930.1 | 4 - 5 | 577 | 138 | |
| GenBank mRNA | AK003135.1 | - | 4 - 5 | 573 | 138 | |
| AK010457.1 | - | 4 - 5 | 578 | 138 | ||
| BC026811.1 | - | 4 - 5 | 557 | 4237838 | 138 | |
| BC055013.1 | - | 4 - 5 | 566 | 5135843 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rdm1
Genomic Map