Assay Details
| Gene Symbol: Cdip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025670.4 | NP_079946.2 | 1 - 2 | 108 | 58 | |
| GenBank mRNA | AK005157.1 | - | 1 - 2 | 94 | 58 | |
| AK017480.1 | - | 1 - 2 | 91 | 58 | ||
| AK031705.1 | - | 1 - 2 | 110 | 58 | ||
| AK051560.1 | - | 1 - 2 | 106 | 58 | ||
| AK149939.1 | - | 1 - 2 | 104 | 58 | ||
| AK150219.1 | - | 1 - 2 | 104 | 58 | ||
| BC004063.1 | - | 1 - 2 | 107 | 3590511 | 58 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK005157.1,AK017480.1,AK031705.1,AK051560.1,AK149939.1,AK150219.1,BC004063.1,NM_025670.4 transcripts.
Set Membership: |
|

Cdip1
Genomic Map