Assay Details
| Gene Symbol: Cdip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025670.4 | NP_079946.2 | 4 - 5 | 432 | 113 | |
| GenBank mRNA | AK005157.1 | - | 4 - 5 | 418 | 113 | |
| AK017480.1 | - | 4 - 5 | 415 | 113 | ||
| AK031705.1 | - | 4 - 5 | 434 | 113 | ||
| AK051560.1 | - | 4 - 5 | 430 | 113 | ||
| AK149939.1 | - | 4 - 5 | 428 | 113 | ||
| AK166141.1 | - | 4 - 5 | 425 | 113 | ||
| BC004063.1 | - | 4 - 5 | 431 | 3590511 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdip1
Genomic Map