Assay Details
| Gene Symbol: Odf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162538.1 | NP_001156010.1 | 14 - 15 | 1768 | 124 | |
| NM_001162539.1 | NP_001156011.1 | 12 - 13 | 1480 | 124 | ||
| NM_025714.4 | NP_079990.2 | 13 - 14 | 1475 | 124 | ||
| GenBank mRNA | AK014630.1 | - | 13 - 14 | 1477 | 124 | |
| AK016726.1 | - | 14 - 15 | 1750 | 124 | ||
| AK220159.1 | - | 3 - 4 | 2952 | 124 | ||
| BC020075.1 | - | 12 - 13 | 1449 | 3979963 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Odf2l
Genomic Map