Assay Details
| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025732.3 | NP_080008.1 | 2 - 3 | 400 | 93 | |
| GenBank mRNA | AK014929.1 | - | 2 - 3 | 401 | 93 | |
| AK017053.1 | - | 2 - 3 | 341 | 93 | ||
| BC052346.1 | - | 2 - 3 | 622 | 6703467 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051052 | AK146023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051052 | BC052346.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051052 | NM_025732.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051052 | AK014929.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121145 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051052 | AK146023.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051052 | BC052346.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051052 | NM_025732.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051052 | AK014929.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158473 | - | - |
| Whole Mouse Genome G4122A | A_51_P189704 | AK014929.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108436 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04604514 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04604513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04148163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04148160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04863443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04604517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04604513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04791717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04791722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04633573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04148162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04244287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04174659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04633567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04148170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04863445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158482 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535500 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535501 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535499 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449395_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535497 | - | - |
| Mouse Genome 430 2.0 Array | 1449395_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449395_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66729_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66729_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500032686.mm.1 | TC0500003565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032686.mm.1 | NM_025732.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032684.mm.1 | TC0500003565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032684.mm.1 | NM_025732.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032683.mm.1 | TC0500003565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032683.mm.1 | NM_025732.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032687.mm.1 | TC0500003565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032687.mm.1 | NM_025732.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032685.mm.1 | TC0500003565.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449395_at | - | - |
| Mouse Expression Set 430 | 1449395_PM_at | - | - |
| Murine Genome U74v2 Set | 168225_i_at | - | - |
| Murine Genome U74v2 Set | 168224_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168225_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168224_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449395_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449395_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449395_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449395_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 851961 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5690484 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218097 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218097 | - | - |
| mouse-6 v1.1 (Illumina) | scl25799.5.1_171-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25799.5.1_171-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690484 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1218097 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218097 | - | - |

Ankrd61
Genomic Map