Assay Details
| Gene Symbol: Aldoart2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277340.1 | NP_001264269.1 | 1 | 88 | 87 | |
| GenBank mRNA | AK006425.1 | - | 1 | 75 | 87 | |
| AK016920.1 | - | 1 | 55 | 87 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK006425.1,AK016920.1,NM_001277340.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063568 | AK016920.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063568 | NR_003959.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063568 | AK006425.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106130 | - | - |
| Whole Mouse Genome G4122A | A_52_P241612 | AK016920.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241612 | EF662062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241612 | NR_003959.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241612 | AK006425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288903 | AK016920.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288903 | NR_003959.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288903 | AK006425.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09823432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10430014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10114458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10114460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10247540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10114461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09823435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10373179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10165536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28452463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09774498 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395826 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423498_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395825 | - | - |
| Mouse Genome 430 2.0 Array | 1423498_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423498_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200003258.mm.1 | NM_001277340.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003258.mm.1 | TC1200000471.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200003259.mm.1 | NM_001277340.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003259.mm.1 | TC1200000471.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423498_at | - | - |
| Mouse Expression Set 430 | 1423498_PM_at | - | - |
| Murine Genome U74v2 Set | 103613_at | - | - |
| Murine Genome U74v2 Set | 103613_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423498_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423498_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423498_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423498_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 817786 | - | - |

Aldoart2
Genomic Map