Assay Details
| Gene Symbol: Fuca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025799.4 | NP_080075.2 | 5 - 6 | 1270 | 105 | |
| GenBank mRNA | AK002675.1 | - | 5 - 6 | 1212 | 105 | |
| AK008653.1 | - | 5 - 6 | 1215 | 105 | ||
| AK017443.1 | - | 5 - 6 | 1240 | 105 | ||
| AK030320.1 | - | 5 - 6 | 1272 | 105 | ||
| AK035607.1 | - | 5 - 6 | 1151 | 105 | ||
| AK146295.1 | - | 5 - 6 | 1199 | 105 | ||
| AK151204.1 | - | 5 - 6 | 1131 | 105 | ||
| AK159288.1 | - | 5 - 6 | 1216 | 105 | ||
| AK163042.1 | - | 3 - 4 | 663 | 105 | ||
| BC004039.1 | - | 5 - 6 | 1221 | 3496187 | 105 | |
| BC018181.1 | - | 5 - 6 | 1230 | 3983067 | 105 | |
| BC094449.1 | - | 5 - 6 | 1185 | 4222249 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fuca2
Genomic Map