Assay Details
| Gene Symbol: Plbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025806.2 | NP_080082.1 | 9 - 10 | 1582 | 109 | |
| GenBank mRNA | AK003169.1 | - | 9 - 10 | 1504 | 109 | |
| AK003711.1 | - | 1 - 2 | 175 | 109 | ||
| AK172021.1 | - | 9 - 10 | 1482 | 109 | ||
| BC019793.1 | - | 9 - 10 | 1534 | 5133074 | 109 | |
More Information
Set Membership: |
|

Plbd1
Genomic Map