Assay Details
| Gene Symbol: Proz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025834.3 | NP_080110.1 | 2 - 3 | 360 | 111 | |
| GenBank mRNA | AK005011.2 | - | 2 - 3 | 304 | 111 | |
| AK008819.1 | - | 2 - 3 | 361 | 111 | ||
| BC038045.1 | - | 2 - 3 | 325 | 4196827 | 111 | |
More Information
Set Membership: |
|

Proz
Genomic Map