Assay Details
| Gene Symbol: Proz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025834.3 | NP_080110.1 | 5 - 6 | 629 | 98 | |
| GenBank mRNA | AK005011.2 | - | 5 - 6 | 573 | 98 | |
| AK008819.1 | - | 5 - 6 | 630 | 98 | ||
| BC024104.1 | - | 5 - 6 | 758 | 5136392 | 98 | |
| BC038045.1 | - | 5 - 6 | 594 | 4196827 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Proz
Genomic Map