Assay Details
| Gene Symbol: Kdelr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025841.4 | NP_080117.1 | 3 - 4 | 486 | 81 | |
| GenBank mRNA | AJ278133.1 | - | 3 - 4 | 479 | 81 | |
| AK003515.1 | - | 3 - 4 | 481 | 81 | ||
| AK007403.1 | - | 3 - 4 | 470 | 81 | ||
| AK150402.1 | - | 3 - 4 | 481 | 81 | ||
| BC007146.1 | - | 3 - 4 | 468 | 3496891 | 81 | |
| CT010232.1 | - | 3 - 4 | 344 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | CT010232.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | AK003515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | AK007403.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | AK150402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | BC007146.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | CT010232.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | AK003515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | AK007403.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | AK150402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P69998 | BC007146.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325244 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | AK007766.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | AK156739.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | AK173287.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | AK217666.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | AY630620.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | BC019994.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | BC034590.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | BC038938.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | BC070463.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | NM_029640.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931118 | NM_180662.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388361 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31121 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | CT010232.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | AK003515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | AK007403.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | AK150402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | BC007146.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | CT010232.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | AK003515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | AK007403.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | AK150402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P69998 | BC007146.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P345080 | AK003515.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P345080 | AK150402.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P345080 | BC007146.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P345080 | NM_025841.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24156416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24156429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24156431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24156430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24156419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24156422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24156417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24156424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24156425 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140755 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137066 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05775039 | - | - |
| Whole Mouse Genome G4122A | A_51_P345080 | AK003515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345080 | AK150402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345080 | BC007146.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345080 | NM_025841.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | CT010232.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | AK003515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | AK007403.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | AK150402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | BC007146.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | CT010232.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | AK003515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | AK007403.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | AK150402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69998 | BC007146.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115223 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137066 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24156517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04552067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04389434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04179941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04481832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04179947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04532285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04742714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04326287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04179948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04475120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04746837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04770392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04416004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04173333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04233735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04876878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04582487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04398430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04179943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04546916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04742713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04409182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04398425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04228185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04914571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04876877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04546913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24156438 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10608708 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417204_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417205_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608708 | - | - |
| Mouse Genome 430 2.0 Array | 1417205_at | - | - |
| Mouse Genome 430 2.0 Array | 1417205_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417204_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417204_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66913_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66913_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500015916.mm.1 | TC0500001732.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500015916.mm.1 | NM_025841.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015916.mm.1 | NM_025841.4 | Refseq |
| Mouse Expression Set 430 | 1417204_at | - | - |
| Mouse Expression Set 430 | 1417205_at | - | - |
| Mouse Expression Set 430 | 1417204_PM_at | - | - |
| Mouse Expression Set 430 | 1417205_PM_at | - | - |
| Murine Genome U74v2 Set | 162201_PM_r_at | - | - |
| Murine Genome U74v2 Set | 101922_PM_at | - | - |
| Murine Genome U74v2 Set | 162201_r_at | - | - |
| Murine Genome U74v2 Set | 101922_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417204_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417205_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417205_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417204_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417204_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417204_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417205_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417205_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 682978 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6900095 | - | - |
| MouseWG-6 v2 Expression BeadChip | 450465 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 450465 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6900095 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000450465 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900095 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2607926 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2761914 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2761914 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2607926 | - | - |
| mouse-6 v1.1 (Illumina) | scl066913.5_18-S | - | - |
| mouse-6 v1.1 (Illumina) | scl27015.4.1_35-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 450465 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900095 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27015.4.1_35-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066913.5_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 450465 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900095 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2607926 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2607926 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2761914 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl27015.4.1_35-S | - | - |

Kdelr2
Genomic Map