Assay Details
| Gene Symbol: Pmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025928.3 | NP_080204.1 | 3 - 4 | 372 | 90 | |
| GenBank mRNA | AF348510.1 | - | 3 - 4 | 382 | 90 | |
| AK011174.1 | - | 3 - 4 | 374 | 90 | ||
| AK012113.1 | - | 3 - 4 | 373 | 90 | ||
| AK013787.1 | - | 3 - 4 | 375 | 90 | ||
| AK078308.1 | - | 3 - 4 | 391 | 90 | ||
| AK168200.1 | - | 3 - 4 | 373 | 90 | ||
| AK180171.1 | - | 1 - 2 | 107 | 90 | ||
| BC034333.1 | - | 3 - 4 | 350 | 90 | ||
| BC037139.1 | - | 3 - 4 | 381 | 4952550 | 90 | |
| BC049223.1 | - | 3 - 4 | 371 | 3257320 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
chromosome segregation
mitotic nuclear division
cell division
regulation of nucleic acid-templated transcription
transcription coactivator activity
protein binding
leucine zipper domain binding
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
chromosome segregation
mitotic nuclear division
cell division
regulation of nucleic acid-templated transcription
transcription coactivator activity
protein binding
leucine zipper domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313620 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313622 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313618 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313617 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438859 | AK011174.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | BC034333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | BC037139.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | BC049223.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | AK011174.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | AK012113.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564742 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24307 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478908 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173719 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103858 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102496 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120961 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438859 | AK011174.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | BC034333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | BC037139.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | BC049223.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | AK011174.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | AK012113.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313615 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04980806 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120961 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00694774 | - | - |
| Whole Mouse Genome G4122A | A_51_P438865 | BC034333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438865 | BC037139.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438865 | BC049223.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438865 | AK011174.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P438865 | AK012113.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123019 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120961 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02368617 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02368607 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313622 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313617 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313620 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03039431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02949287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03084230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03084227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02566831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02916118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02728597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02595535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02778967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02983286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313610 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499369 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499368 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416707_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438173_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499366 | - | - |
| Mouse Genome 430 2.0 Array | 1438173_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1416707_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416707_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1438173_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67037_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67037_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300017509.mm.1 | NM_025928.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017509.mm.1 | TC0300002310.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416707_a_at | - | - |
| Mouse Expression Set 430 | 1416707_PM_a_at | - | - |
| Mouse Expression Set 430 | 1438173_PM_x_at | - | - |
| Mouse Expression Set 430 | 1438173_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438173_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416707_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438173_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416707_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416707_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438173_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438173_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416707_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 543499 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5130273 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5130273 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005130273 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2711410 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2711410 | - | - |
| mouse-6 v1.1 (Illumina) | scl067037.1_35-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5130273 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067037.1_35-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2711410 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2711410 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067037.1_35-S | - | - |

Pmf1
Genomic Map