Assay Details
| Gene Symbol: Magt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190409.1 | NP_001177338.1 | 3 - 4 | 527 | 77 | |
| NM_025952.4 | NP_080228.4 | 3 - 4 | 527 | 77 | ||
| GenBank mRNA | AK010320.1 | - | 3 - 4 | 463 | 77 | |
| AK012185.1 | - | 3 - 4 | 404 | 77 | ||
| AK013243.1 | - | 3 - 4 | 408 | 77 | ||
| AK018623.1 | - | 3 - 4 | 430 | 77 | ||
| AK136622.1 | - | 3 - 4 | 409 | 77 | ||
| AK144696.1 | - | 3 - 4 | 406 | 77 | ||
| BC003881.1 | - | 3 - 4 | 414 | 3496572 | 77 | |
| DQ000005.1 | - | 3 - 4 | 527 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Magt1
Genomic Map