Assay Details
| Gene Symbol: Cers4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026058.4 | NP_080334.3 | 8 - 9 | 928 | 65 | |
| GenBank mRNA | AK013554.1 | - | 8 - 9 | 880 | 65 | |
| AK021316.1 | - | 1 - 2 | 35 | 65 | ||
| AK028937.1 | - | 8 - 9 | 854 | 65 | ||
| AK028945.1 | - | 8 - 9 | 831 | 65 | ||
| AK030280.1 | - | 8 - 9 | 868 | 65 | ||
| AK134887.1 | - | 7 - 8 | 1587 | 65 | ||
| AY029531.1 | - | 8 - 9 | 829 | 65 | ||
| BC003946.1 | - | 8 - 9 | 815 | 3487343 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cers4
Genomic Map