Assay Details
| Gene Symbol: Lyzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026092.3 | NP_080368.1 | 2 - 3 | 281 | 83 | |
| GenBank mRNA | AK006241.1 | - | 2 - 3 | 282 | 83 | |
| AK006630.1 | - | 2 - 3 | 282 | 83 | ||
| AY572447.1 | - | 2 - 3 | 240 | 83 | ||
| BC048450.1 | - | 2 - 3 | 252 | 6774504 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lyzl1
Genomic Map