Assay Details
Gene Symbol: Tmed11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026109.2 | NP_080385.2 | 4 - 5 | 546 | 107 | |
GenBank mRNA | AK007381.1 | - | 4 - 5 | 555 | 107 | |
AK019032.1 | - | 4 - 5 | 535 | 107 | ||
BC132664.1 | - | 4 - 5 | 518 | 40130941 | 107 | |
BC132666.1 | - | 4 - 5 | 518 | 40130943 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974101 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974103 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974100 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974102 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974099 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | BY707479.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | NM_026109.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | AK007381.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | AK019032.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | BC132664.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | BY707479.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | NM_026109.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | AK007381.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | AK019032.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146741 | BC132664.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961023 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849528 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694703 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214644 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341689 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740220 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180808 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169907 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125560 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | BY707479.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | NM_026109.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | AK007381.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | AK019032.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | BC132664.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | BY707479.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | NM_026109.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | AK007381.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | AK019032.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146741 | BC132664.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | BY707479.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | NM_026109.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | AK007381.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | AK019032.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | BC132664.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | BY707479.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | NM_026109.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | AK007381.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | AK019032.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P146741 | BC132664.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23974198 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169907 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01184623 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125560 | - | - |
Whole Mouse Genome G4122A | A_51_P146741 | BY707479.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P146741 | NM_026109.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P146741 | AK007381.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P146741 | AK019032.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P146741 | BC132664.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P146741 | BY707479.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P146741 | NM_026109.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P146741 | AK007381.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P146741 | AK019032.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P146741 | BC132664.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133902 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169907 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01184623 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125560 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974101 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974099 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974102 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974103 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23974100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23974169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974077 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974076 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974162 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974111 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974124 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974171 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04690033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04209451 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974135 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974187 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974097 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974073 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974133 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974068 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974067 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04244113 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04244128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974094 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23974070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04690036 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10532264 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532262 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532266 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532263 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532265 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418111_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10532261 | - | - |
Mouse Genome 430 2.0 Array | 1418111_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418111_at | - | - |
Mouse Genome 430 2.0 Array | 1445960_at | - | - |
Mouse Genome 430 2.0 Array | 1445960_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67366_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67366_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500026186.mm.1 | TC0500002956.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500026186.mm.1 | NM_026109.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500026186.mm.1 | NM_026109.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500026187.mm.1 | TC0500002956.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500026187.mm.1 | NM_026109.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500026187.mm.1 | NM_026109.2 | Refseq |
Mouse Expression Set 430 | 1418111_at | - | - |
Mouse Expression Set 430 | 1418111_PM_at | - | - |
Murine Genome U74v2 Set | 165490_PM_r_at | - | - |
Murine Genome U74v2 Set | 165490_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1418111_at | - | - |
Mouse Genome 430A 2.0 Array | 1418111_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418111_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418111_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 387469 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4610592 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004610592 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2632811 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2632811 | - | - |
mouse-6 v1.1 (Illumina) | scl26243.5.1_124-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4610592 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl26243.5.1_124-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610592 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2632811 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2632811 | - | - |
