Assay Details
| Gene Symbol: Paxbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026110.2 | NP_080386.3 | 12 - 13 | 2093 | 74 | |
| GenBank mRNA | AK007365.1 | - | 6 - 7 | 798 | 74 | |
| AK011477.1 | - | 2 - 3 | 177 | 74 | ||
| AY033907.1 | - | 12 - 13 | 1878 | 74 | ||
| AY033908.1 | - | 12 - 13 | 1895 | 74 | ||
| BC014838.1 | - | 5 - 6 | 578 | 3588322 | 74 | |
| BC027145.1 | - | 4 - 5 | 513 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Paxbp1
Genomic Map