| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300013043.mm.1 |
NM_001114087.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013043.mm.1 |
NM_001114088.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013043.mm.1 |
NM_026131.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013043.mm.1 |
NR_104287.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013043.mm.1 |
TC1300002061.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013046.mm.1 |
NM_001114087.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013046.mm.1 |
NM_001114088.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013046.mm.1 |
NM_026131.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013046.mm.1 |
NR_104287.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013046.mm.1 |
TC1300002061.mm.1 |
MTA TC |