Assay Details
| Gene Symbol: Ceacam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007543.4 | NP_031569.1 | 4 - 5 | 1058 | 66 | |
| GenBank mRNA | AB500065.1 | - | 4 - 5 | 883 | 66 | |
| BC024320.1 | - | 4 - 5 | 867 | 4241449 | 66 | |
| X76085.1 | - | 4 - 5 | 916 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2288983 | AB500065.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2288983 | AF101164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2288983 | BC024320.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2288983 | X76085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2288983 | NM_001113369.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2386236 | BC016891.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565362 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364265 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371288 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758714 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79623 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177371 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2288983 | AB500065.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2288983 | AF101164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2288983 | BC024320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2288983 | X76085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2288983 | NM_001113369.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2386236 | BC016891.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P171728 | BC024320.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P171728 | NM_001113369.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P171728 | NM_007543.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P171728 | NM_001113368.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P171728 | X67282.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010731 | - | - |
| Whole Mouse Genome G4122A | A_52_P9671 | AB500065.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9671 | AF101164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9671 | BC024320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9671 | X76085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P9671 | NM_001113369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240706 | BC024320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240706 | NM_001113369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240706 | NM_007543.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240706 | NM_001113368.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240706 | BC121189.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134349 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114987 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06128308 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06128310 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06128307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06128307 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561056 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561057 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561058 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421603_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561055 | - | - |
| Mouse Genome 430 2.0 Array | 1422123_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1460681_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460681_at | - | - |
| Mouse Genome 430 2.0 Array | 1460682_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422123_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1460682_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1421603_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421603_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26367_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26367_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022783.mm.1 | TC0700002636.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022783.mm.1 | NM_001113368.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022783.mm.1 | NM_001113368.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022783.mm.1 | NM_001113369.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022783.mm.1 | NM_001113369.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022783.mm.1 | NM_007543.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022783.mm.1 | NM_007543.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022780.mm.1 | TC0700002636.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022780.mm.1 | NM_001113368.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022780.mm.1 | NM_001113368.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022780.mm.1 | NM_001113369.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022780.mm.1 | NM_001113369.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022780.mm.1 | NM_007543.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022780.mm.1 | NM_007543.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022779.mm.1 | TC0700002636.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022779.mm.1 | NM_001113368.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022779.mm.1 | NM_001113368.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022779.mm.1 | NM_001113369.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022779.mm.1 | NM_001113369.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022779.mm.1 | NM_007543.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022779.mm.1 | NM_007543.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022781.mm.1 | TC0700002636.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022781.mm.1 | NM_001113368.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022781.mm.1 | NM_001113368.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022781.mm.1 | NM_001113369.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022781.mm.1 | NM_001113369.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022781.mm.1 | NM_007543.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022781.mm.1 | NM_007543.4 | Refseq |
| Mouse Expression Set 430 | 1422123_s_at | - | - |
| Mouse Expression Set 430 | 1421603_a_at | - | - |
| Mouse Expression Set 430 | 1460682_PM_s_at | - | - |
| Mouse Expression Set 430 | 1422123_PM_s_at | - | - |
| Mouse Expression Set 430 | 1460682_s_at | - | - |
| Mouse Expression Set 430 | 1460681_at | - | - |
| Mouse Expression Set 430 | 1421603_PM_a_at | - | - |
| Mouse Expression Set 430 | 1460681_PM_at | - | - |
| Murine Genome U74v2 Set | 101907_PM_s_at | - | - |
| Murine Genome U74v2 Set | 102804_PM_at | - | - |
| Murine Genome U74v2 Set | 101908_PM_s_at | - | - |
| Murine Genome U74v2 Set | 101907_s_at | - | - |
| Murine Genome U74v2 Set | 102804_at | - | - |
| Murine Genome U74v2 Set | 101908_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460682_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460681_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460681_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460682_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421603_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421603_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422123_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422123_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421603_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421603_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 397624 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6020133 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2970292 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2970292 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6020133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006020133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002970292 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2597854 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2597854 | - | - |
| mouse-6 v1.1 (Illumina) | scl0026367.1_126-S | - | - |
| mouse-6 v1.1 (Illumina) | scl026367.4_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2970292 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6020133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl026367.4_9-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0026367.1_126-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2970292 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2597854 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239472 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2597854 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0026367.1_126-S | - | - |

Ceacam2
Genomic Map