Assay Details
| Gene Symbol: Cap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154002.1 | - | 1 - 2 | 244 | 105 | |
| AK158150.1 | - | 1 - 2 | 54 | 105 | ||
| L12367.1 | - | 1 - 2 | 105 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154002.1, L12367.1 transcripts.
Set Membership: |
|

Cap1
Genomic Map