Assay Details
| Gene Symbol: Cap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301067.1 | NP_001287996.1 | 4 - 5 | 544 | 71 | |
| NM_007598.4 | NP_031624.2 | 4 - 5 | 528 | 71 | ||
| GenBank mRNA | AK053351.1 | - | 4 - 5 | 352 | 71 | |
| AK137222.1 | - | 4 - 5 | 333 | 71 | ||
| AK144094.1 | - | 4 - 5 | 336 | 71 | ||
| AK154002.1 | - | 4 - 5 | 548 | 71 | ||
| AK156556.1 | - | 4 - 5 | 362 | 71 | ||
| AK158150.1 | - | 3 - 4 | 1192 | 71 | ||
| AK165565.1 | - | 4 - 5 | 331 | 71 | ||
| AK169652.1 | - | 4 - 5 | 323 | 71 | ||
| AK170901.1 | - | 4 - 5 | 339 | 71 | ||
| AK171388.1 | - | 4 - 5 | 346 | 71 | ||
| BC005446.1 | - | 3 - 4 | 327 | 3600514 | 71 | |
| BC005472.1 | - | 3 - 4 | 320 | 3590890 | 71 | |
| L12367.1 | - | 4 - 5 | 354 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cap1
Genomic Map