Assay Details
| Gene Symbol: Capn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110504.1 | NP_001103974.1 | 14 - 15 | 1689 | 90 | |
| NM_007600.3 | NP_031626.1 | 14 - 15 | 1741 | 90 | ||
| GenBank mRNA | AF021847.1 | - | 14 - 15 | 1620 | 90 | |
| AF084459.1 | - | 13 - 14 | 1607 | 90 | ||
| AK088547.1 | - | 14 - 15 | 1661 | 90 | ||
| AK149090.1 | - | 10 - 11 | 1089 | 90 | ||
| AK149706.1 | - | 14 - 15 | 1682 | 90 | ||
| AK150084.1 | - | 14 - 15 | 1680 | 90 | ||
| AK160940.1 | - | 14 - 15 | 1681 | 90 | ||
| AK165258.1 | - | 14 - 15 | 1701 | 90 | ||
| AK168063.1 | - | 14 - 15 | 1680 | 90 | ||
| AK171405.1 | - | 14 - 15 | 1644 | 90 | ||
| BC026138.1 | - | 14 - 15 | 1656 | 4218148 | 90 | |
| BC050276.1 | - | 14 - 15 | 1657 | 4923347 | 90 | |
More Information
Set Membership: |
|

Capn1
Genomic Map