Assay Details
| Gene Symbol: Capn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110504.1 | NP_001103974.1 | 17 - 18 | 1881 | 59 | |
| NM_007600.3 | NP_031626.1 | 17 - 18 | 1933 | 59 | ||
| GenBank mRNA | AF021847.1 | - | 17 - 18 | 1812 | 59 | |
| AF084459.1 | - | 16 - 17 | 1799 | 59 | ||
| AK088547.1 | - | 17 - 18 | 1853 | 59 | ||
| AK149090.1 | - | 13 - 14 | 1281 | 59 | ||
| AK149706.1 | - | 17 - 18 | 1874 | 59 | ||
| AK150084.1 | - | 17 - 18 | 1872 | 59 | ||
| AK160940.1 | - | 17 - 18 | 1873 | 59 | ||
| AK165258.1 | - | 17 - 18 | 1893 | 59 | ||
| AK168063.1 | - | 17 - 18 | 1872 | 59 | ||
| AK171405.1 | - | 17 - 18 | 1836 | 59 | ||
| BC026138.1 | - | 17 - 18 | 1848 | 4218148 | 59 | |
| BC050276.1 | - | 17 - 18 | 1849 | 4923347 | 59 | |
More Information
Set Membership: |
|

Capn1
Genomic Map