Assay Details
| Gene Symbol: Capza2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007604.2 | NP_031630.1 | 3 - 4 | 179 | 124 | |
| GenBank mRNA | AK007550.1 | - | 3 - 4 | 178 | 124 | |
| AK008204.1 | - | 3 - 4 | 165 | 124 | ||
| AK150587.1 | - | 3 - 4 | 181 | 124 | ||
| AK150751.1 | - | 3 - 4 | 246 | 124 | ||
| AY544984.1 | - | 3 - 4 | 175 | 124 | ||
| BC082589.1 | - | 3 - 4 | 179 | 30626416 | 124 | |
| U16741.1 | - | 3 - 4 | 260 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capza2
Genomic Map