Assay Details
| Gene Symbol: Car3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007606.3 | NP_031632.2 | 2 - 3 | 267 | 95 | |
| GenBank mRNA | AK002349.1 | - | 2 - 3 | 294 | 95 | |
| AK003671.1 | - | 2 - 3 | 297 | 95 | ||
| BC011129.1 | - | 2 - 3 | 267 | 4195712 | 95 | |
| M27796.1 | - | 2 - 3 | 316 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car3
Genomic Map