Assay Details
| Gene Symbol: Ctnnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085448.1 | NP_001078917.1 | 4 - 5 | 845 | 71 | |
| NM_001085449.1 | NP_001078918.1 | 3 - 4 | 556 | 71 | ||
| NM_001085450.1 | NP_001078919.1 | 4 - 5 | 845 | 71 | ||
| NM_001085453.1 | NP_001078922.1 | 4 - 5 | 615 | 71 | ||
| NM_007615.4 | NP_031641.2 | 5 - 6 | 963 | 71 | ||
| GenBank mRNA | AB093237.1 | - | 5 - 6 | 679 | 71 | |
| AK133193.1 | - | 4 - 5 | 566 | 71 | ||
| AK161790.1 | - | 5 - 6 | 739 | 71 | ||
| AK164485.1 | - | 4 - 5 | 830 | 71 | ||
| AK168453.1 | - | 3 - 4 | 555 | 70 | ||
| AK195355.1 | - | 2 - 3 | 215 | 71 | ||
| BC043108.1 | - | 4 - 5 | 780 | 6406902 | 71 | |
| BC046589.1 | - | 3 - 4 | 356 | 71 | ||
| BC054544.1 | - | 5 - 6 | 896 | 6408956 | 71 | |
| Z17804.1 | - | 758 | 71 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnd1
Genomic Map