Assay Details
| Gene Symbol: Ctnnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085448.1 | NP_001078917.1 | 6 - 7 | 1852 | 93 | |
| NM_001085449.1 | NP_001078918.1 | 5 - 6 | 1563 | 93 | ||
| NM_001085450.1 | NP_001078919.1 | 6 - 7 | 1852 | 93 | ||
| NM_001085453.1 | NP_001078922.1 | 6 - 7 | 1622 | 93 | ||
| NM_007615.4 | NP_031641.2 | 7 - 8 | 1970 | 93 | ||
| GenBank mRNA | AB093237.1 | - | 7 - 8 | 1686 | 93 | |
| AK133193.1 | - | 6 - 7 | 1573 | 93 | ||
| AK148871.1 | - | 2 - 3 | 633 | 93 | ||
| AK161790.1 | - | 7 - 8 | 1746 | 93 | ||
| AK164485.1 | - | 6 - 7 | 1837 | 93 | ||
| AK168453.1 | - | 5 - 6 | 1561 | 93 | ||
| AK206736.1 | - | 1 - 2 | 197 | 93 | ||
| BC043108.1 | - | 6 - 7 | 1787 | 6406902 | 93 | |
| BC046589.1 | - | 5 - 6 | 1363 | 93 | ||
| BC054544.1 | - | 7 - 8 | 1903 | 6408956 | 93 | |
| Z17804.1 | - | 1765 | 93 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnd1
Genomic Map