Assay Details
| Gene Symbol: Cbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007623.2 | NP_031649.2 | 1 - 2 | 158 | 61 | |
| GenBank mRNA | AK157869.1 | - | 1 - 2 | 134 | 61 | |
| BC035199.1 | - | 1 - 2 | 158 | 5137505 | 61 | |
| X62537.1 | - | 1 - 2 | 225 | 61 | ||
| Y13274.1 | - | 1 - 2 | 82 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cbx2
Genomic Map