Assay Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007661.3 | NP_031687.2 | 10 - 11 | 1166 | 67 | |
| GenBank mRNA | AK077668.1 | - | 10 - 11 | 1167 | 67 | |
| AK147133.1 | - | 10 - 11 | 1144 | 67 | ||
| BC052920.1 | - | 10 - 11 | 1244 | 6489832 | 67 | |
| L37092.1 | - | 9 - 10 | 1738 | 67 | ||
| M58633.1 | - | 5 - 6 | 514 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk11b
Genomic Map