Assay Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007661.3 | NP_031687.2 | 17 - 18 | 2008 | 75 | |
| GenBank mRNA | AK077668.1 | - | 17 - 18 | 2009 | 75 | |
| AK147133.1 | - | 17 - 18 | 1986 | 75 | ||
| BC025058.1 | - | 6 - 7 | 658 | 75 | ||
| BC052920.1 | - | 17 - 18 | 2086 | 6489832 | 75 | |
| L37092.1 | - | 16 - 17 | 2580 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk11b
Genomic Map