Assay Details
| Gene Symbol: Cenpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA016357.1 | - | 1 - 2 | 157 | 85 | |
| AF012709.1 | - | 1 - 2 | 213 | 85 | ||
| AK010718.1 | - | 1 - 2 | 175 | 85 | ||
| AK011399.1 | - | 1 - 2 | 158 | 85 | ||
| AK053763.1 | - | 1 - 2 | 182 | 85 | ||
| AK089033.1 | - | 1 - 2 | 186 | 85 | ||
| BC011038.1 | - | 1 - 2 | 193 | 4018429 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573890 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573891 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573889 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121459 | AK089033.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121459 | BC011038.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121459 | BC012280.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121459 | AK040313.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121459 | AK041138.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490266 | AK084665.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490266 | AK089185.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490266 | BC058545.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927784 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147568 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139356 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103446 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121459 | AK089033.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121459 | BC011038.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121459 | BC012280.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121459 | AK040313.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121459 | AK041138.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273979 | BC011038.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273979 | AK053763.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273979 | NM_007681.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273979 | AF012709.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273979 | AK011399.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23573888 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139356 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103446 | - | - |
| Whole Mouse Genome G4122A | A_52_P328630 | - | - |
| Whole Mouse Genome G4122A | A_51_P273979 | BC011038.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273979 | AK053763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273979 | NM_007681.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273979 | AF012709.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273979 | AK011399.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113336 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121459 | AK089033.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121459 | BC011038.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121459 | BC012280.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121459 | AK040313.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121459 | AK041138.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139356 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103446 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04723858 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04723864 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04723863 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04723855 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04723865 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04723859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573892 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573891 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23573890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23573947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04824501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04476216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04979941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04373727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04386671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04476205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04626989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04723865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23573869 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10520523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520522 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450842_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444416_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441864_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10520521 | - | - |
| Mouse Genome 430 2.0 Array | 1450842_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1441864_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1444416_at | - | - |
| Mouse Genome 430 2.0 Array | 1450842_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1444416_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441864_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12615_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12615_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500002330.mm.1 | TC0500000298.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500002330.mm.1 | NM_007681.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002331.mm.1 | TC0500000298.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500002331.mm.1 | NM_007681.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002338.mm.1 | TC0500000298.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500002338.mm.1 | NM_007681.3 | Refseq |
| Mouse Expression Set 430 | 1450842_PM_a_at | - | - |
| Mouse Expression Set 430 | 1450842_a_at | - | - |
| Mouse Expression Set 430 | 1441864_PM_x_at | - | - |
| Mouse Expression Set 430 | 1441864_x_at | - | - |
| Mouse Expression Set 430 | 1444416_at | - | - |
| Mouse Expression Set 430 | 1444416_PM_at | - | - |
| Murine Genome U74v2 Set | 100616_at | - | - |
| Murine Genome U74v2 Set | 100616_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450842_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450842_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450842_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450842_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444416_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441864_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444416_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441864_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 913765 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103840358 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5080154 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103840358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005080154 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236574 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236574 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2471161 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003955.1_3-S | - | - |
| mouse-6 v1.1 (Illumina) | scl012615.5_26-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5080154 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103840358 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012615.5_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080154 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103840358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1236574 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2471161 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236574 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl012615.5_26-S | - | - |

Cenpa
Genomic Map