Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007704.2 | NP_031730.1 | 16 - 17 | 2052 | 92 | |
| NM_172696.2 | NP_766284.2 | 42 - 43 | 5502 | 92 | ||
| GenBank mRNA | AF060539.1 | - | 16 - 17 | 1952 | 92 | |
| AK077268.1 | - | 29 - 30 | 3784 | 92 | ||
| AV287690.2 | - | 1 - 2 | 113 | 92 | ||
| BC057124.1 | - | 16 - 17 | 1731 | 92 | ||
| BC082787.1 | - | 31 - 32 | 3895 | 92 | ||
| BC141404.1 | - | 42 - 43 | 5478 | 9056017 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inadl
Genomic Map