Assay Details
| Gene Symbol: Coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198835.1 | NP_001185764.1 | 1 - 2 | 64 | 61 | |
| GenBank mRNA | AF006741.1 | - | 1 - 2 | 48 | 61 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF006741.1,NM_001198835.1 transcripts.
Set Membership: |
|

Coch
Genomic Map