Assay Details
| Gene Symbol: Coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198835.1 | NP_001185764.1 | 4 - 5 | 327 | 92 | |
| NM_007728.5 | NP_031754.1 | 3 - 4 | 526 | 92 | ||
| GenBank mRNA | AF006741.1 | - | 4 - 5 | 311 | 92 | |
| AK028690.1 | - | 3 - 4 | 413 | 92 | ||
| AK137336.1 | - | 3 - 4 | 528 | 92 | ||
| AK156430.1 | - | 4 - 5 | 296 | 92 | ||
| AK171875.1 | - | 4 - 5 | 296 | 92 | ||
| BC045137.1 | - | 4 - 5 | 278 | 6306601 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coch
Genomic Map