Assay Details
| Gene Symbol: Coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198835.1 | NP_001185764.1 | 6 - 7 | 524 | 74 | |
| NM_007728.5 | NP_031754.1 | 5 - 6 | 723 | 74 | ||
| GenBank mRNA | AF006741.1 | - | 6 - 7 | 508 | 74 | |
| AK028690.1 | - | 5 - 6 | 610 | 74 | ||
| AK137336.1 | - | 5 - 6 | 725 | 74 | ||
| AK156430.1 | - | 6 - 7 | 493 | 74 | ||
| AK171875.1 | - | 6 - 7 | 493 | 74 | ||
| BC045137.1 | - | 6 - 7 | 475 | 6306601 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coch
Genomic Map