Assay Details
| Gene Symbol: Coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198835.1 | NP_001185764.1 | 10 - 11 | 1052 | 79 | |
| NM_007728.5 | NP_031754.1 | 9 - 10 | 1251 | 79 | ||
| GenBank mRNA | AF006741.1 | - | 10 - 11 | 1036 | 79 | |
| AK028690.1 | - | 9 - 10 | 1138 | 79 | ||
| AK137336.1 | - | 9 - 10 | 1253 | 79 | ||
| AK156430.1 | - | 10 - 11 | 1021 | 79 | ||
| AK171875.1 | - | 10 - 11 | 1021 | 79 | ||
| BC045137.1 | - | 10 - 11 | 1003 | 6306601 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coch
Genomic Map