Assay Details
| Gene Symbol: Cpeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252525.1 | NP_001239454.1 | 4 - 5 | 714 | 125 | |
| NM_001252526.1 | NP_001239455.1 | 4 - 5 | 711 | 125 | ||
| NM_007755.5 | NP_031781.1 | 4 - 5 | 711 | 125 | ||
| GenBank mRNA | AK136088.1 | - | 4 - 5 | 714 | 125 | |
| BC125476.1 | - | 4 - 5 | 631 | 40129991 | 125 | |
| BC144948.1 | - | 4 - 5 | 628 | 125 | ||
| Y08260.1 | - | 4 - 5 | 630 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpeb1
Genomic Map