Assay Details
| Gene Symbol: Ncan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007789.3 | NP_031815.2 | 4 - 5 | 772 | 56 | |
| GenBank mRNA | BC065118.1 | - | 4 - 5 | 738 | 6853253 | 56 |
| X84727.1 | - | 4 - 5 | 734 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ncan
Genomic Map