Assay Details
| Gene Symbol: Ncan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007789.3 | NP_031815.2 | 5 - 6 | 896 | 62 | |
| GenBank mRNA | BC065118.1 | - | 5 - 6 | 862 | 6853253 | 62 |
| X84727.1 | - | 5 - 6 | 858 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ncan
Genomic Map