Assay Details
| Gene Symbol: Ncan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007789.3 | NP_031815.2 | 6 - 7 | 1194 | 69 | |
| GenBank mRNA | BC065118.1 | - | 6 - 7 | 1160 | 6853253 | 69 |
| X84727.1 | - | 6 - 7 | 1156 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ncan
Genomic Map