Assay Details
| Gene Symbol: Ctsk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007802.4 | NP_031828.2 | 2 - 3 | 241 | 67 | |
| GenBank mRNA | AK003425.1 | - | 2 - 3 | 166 | 67 | |
| AK132648.1 | - | 2 - 3 | 171 | 67 | ||
| BC046320.1 | - | 2 - 3 | 195 | 6390854 | 67 | |
| X94444.1 | - | 1 - 2 | 113 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsk
Genomic Map