Assay Details
| Gene Symbol: Cyp19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007810.3 | NP_031836.1 | 2 - 3 | 340 | 72 | |
| GenBank mRNA | AK143323.1 | - | 2 - 3 | 420 | 72 | |
| BC103670.1 | - | 2 - 3 | 323 | 40047093 | 72 | |
| BC104489.1 | - | 2 - 3 | 323 | 40047095 | 72 | |
| D00659.1 | - | 2 - 3 | 345 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp19a1
Genomic Map