| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400026405.mm.1 |
TC0400003318.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026405.mm.1 |
NM_007822.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026405.mm.1 |
NM_007822.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026406.mm.1 |
TC0400003318.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026406.mm.1 |
NM_007822.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026406.mm.1 |
NM_007822.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026407.mm.1 |
TC0400003318.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026407.mm.1 |
NM_007822.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026407.mm.1 |
NM_007822.2 |
Refseq |