Assay Details
| Gene Symbol: Cyp4a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007822.2 | NP_031848.1 | 10 - 11 | 1306 | 66 | |
| GenBank mRNA | AK098088.1 | - | 10 - 11 | 1303 | 66 | |
| AK149392.1 | - | 10 - 11 | 1304 | 66 | ||
| AK165452.1 | - | 10 - 11 | 1304 | 66 | ||
| BC089609.1 | - | 10 - 11 | 1306 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
oxidation-reduction process
monooxygenase activity
fatty acid binding
iron ion binding
arachidonic acid monooxygenase activity
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
alkane 1-monooxygenase activity
heme binding
metal ion binding
arachidonic acid binding
arachidonic acid omega-hydroxylase activity
oxidation-reduction process
monooxygenase activity
fatty acid binding
iron ion binding
arachidonic acid monooxygenase activity
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
alkane 1-monooxygenase activity
heme binding
metal ion binding
arachidonic acid binding
arachidonic acid omega-hydroxylase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P238576 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66794 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65159 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138461 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P238576 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P238576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23224695 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05366570 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138461 | - | - |
| Whole Mouse Genome G4122A | A_51_P238576 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118338 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118978 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05366570 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138461 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822927 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822928 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822920 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822929 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822933 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822923 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23224691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03336817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03403937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03336828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03712408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03202140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03644911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03466747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03562317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03562321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04038630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03336839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03336829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23224689 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515196 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515194 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515197 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515191 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515198 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515200 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423257_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515187 | - | - |
| Mouse Genome 430 2.0 Array | 1423257_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423257_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13119_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13119_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400026401.mm.1 | TC0400003318.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026401.mm.1 | NM_007822.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026401.mm.1 | NM_007822.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026402.mm.1 | TC0400003318.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026402.mm.1 | NM_007822.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026402.mm.1 | NM_007822.2 | Refseq |
| Mouse Expression Set 430 | 1423257_at | - | - |
| Mouse Expression Set 430 | 1423257_PM_at | - | - |
| Murine Genome U74v2 Set | 101103_PM_at | - | - |
| Murine Genome U74v2 Set | 101103_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423257_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423257_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423257_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423257_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 479744 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2370110 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2370110 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002370110 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2691680 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2691680 | - | - |
| mouse-6 v1.1 (Illumina) | scl23970.12_438-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2370110 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23970.12_438-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370110 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2691680 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2691680 | - | - |

Cyp4a14
Genomic Map